Titanium Powder, -325 mesh, 99% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1038636

Quantity 500g

 View more versions of this product

Catalog No. AA1038636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-32-6
    Physical Form Powder
    Assay Percent Range 99% (Metals basis)
    MDL Number MFCD00011264
    Chemical Name or Material Titanium

    Specifications

    Quantity 500g
    Color Dark gray
    DOT Information DOT Class: 4.2, Packaging Group: II
    EINECS Number 231-142-3
    Physical Form Powder
    Assay Percent Range 99% (Metals basis)
    MDL Number MFCD00011264
    CAS 7440-32-6
    Odor Odorless
    Health Hazard 1 H251; Self-heating: may catch fire.
    Appearance Dark Gray powder
    Chemical Name or Material Titanium

    H251; Self-heating: may catch fire.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit