Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Titanium sponge, 3-19mm (0.12-0.75in), 99.95% (metals basis)
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Gray |
| Quantity | 50 g |
| Odor | Odorless |
| Form | Sponge |
| UN Number | UN2878 |
| Percent Purity | 99.95% |
| Assay | (metals basis) |
| Chemical Name or Material | Titanium sponge, 3 to 19mm (0.12 to 0.75 in.) |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H228 Flammable solid. |
| Precautionary Statement | GHS P Statement P210-P280-P240-P241-P370+P378a Keep away from heat/sparks/open flames/hot surfaces. No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Ground/bond container and receiving equipment. Use explosion-proof electrical/ventilating/lighting/ equipment. In case of fire: Use for extinction: CO2, powder or water spray. |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: TITANIUM SPONGE GRANULES |
| EINECSNumber | 231-142-3 |
| RTECSNumber | XR1700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction