Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TJAP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP238152 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP238152 0.1 mL
NB404682 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP238152 Supplier Novus Biologicals Supplier No. NBP238152
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TJAP1 Polyclonal specifically detects TJAP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TJAP1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q5JTD0
Gene Alias DKFZp686F06131, PILTTJP4tight junction protein 4 (peripheral), Protein incorporated later into tight junctions, tight junction associated protein 1 (peripheral), Tight junction protein 4, tight junction-associated protein 1
Gene Symbols TJAP1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: SASSEEDLLVSWQRAFVDRTPPPAAVAQRTAFGRDALPELQRHFAHSPADRDEVVQAPSARPEESELLLPTEP
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93643
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.