Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TK1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501559970
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9970 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9970 Supplier Bethyl Laboratories, Inc Supplier No. A301280AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Thymidine kinase (TK) belongs to a group of enzymes such as dihydrofolate reductase, thymidylate synthase, and DNA polymerase that are involved in DNA synthesis and precursor production. High levels of these enzymes are present in proliferating cells, and low levels are found in resting cells, due to multiple regulatory mechanisms that ensure exclusive expression of these enzymes in replicating cells. TK is responsible for catalyzing the phosphorylation of thymidine, which functions as a part of the pyrimidine salvage pathway involved in DNA synthesis. The activities of enzymes such as TK are essential for the activation of several chemotherapeutically important nucleoside analogues that are administered as prodrugs.

Specifications

Antigen TK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TK1
Gene Accession No. P04183
Gene Alias TK2
Gene Symbols TK1
Host Species Rabbit
Immunogen Between 205 and 234
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7083
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.