Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TL1A/TNFSF15 Antibody (2116C), Alexa Fluor™ 594, R&D Systems™
SDP

Catalog No. MB7442AF594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MB7442AF594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MB7442AF594 Supplier Novus Biologicals Supplier No. MAB7442AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Human samples. It is validated for Western Blot.

Specifications

Antigen TL1A/TNFSF15
Applications Western Blot
Classification Monoclonal
Clone 2116C
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1
Gene Symbols TNFSF15
Host Species Rabbit
Immunogen Partial recombinant human TL1A protein (amino acids 72-251) [UniProt O95150]
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Biologically Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9966.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.