Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TL1A/TNFSF15 Antibody (VEGI/2052R), FITC, Novus Biologicals™
SDP

Catalog No. NB390537 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390537 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390537 Supplier Novus Biologicals Supplier No. NBP308802F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TL1A/TNFSF15 Monoclonal antibody specifically detects TL1A/TNFSF15 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen TL1A/TNFSF15
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone VEGI/2052R
Conjugate FITC
Formulation PBS
Gene Alias MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1
Host Species Rabbit
Immunogen Recombinant full-length human TL1A/TNFSF15 protein (Uniprot: O95150)
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9966
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.