Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ TL4-12

Click to view available options
Quantity:
10 mg
50 mg
Description
TL4-12 is a potent MAP4K2 (GCK) inhibitor (IC50 = 37 nM). Exhibits >70-fold selectivity for MAP4K2 over TAK1 (IC50 = 2.7 μM for TAK1). Inhibits IL-1 and TGFβ-induced p38 MAPK phosphorylation, in vitro.
Specifications
Specifications
Chemical Name or Material | 4-Methyl-3-[[6-(methylamino)-4-pyrimidinyl]oxy]-N-[3-(4-methyl-1-piperazinyl)-5-(trifluoromethyl)phenyl]benzamide |
CAS | 1620820-12-3 |
Quantity | 10 mg |
Molecular Formula | C25H27F3N6O2 |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction