Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLE1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP204105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP204105 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP204105 Supplier Novus Biologicals Supplier No. NBP204105
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TLE1 Polyclonal specifically detects TLE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen TLE1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:500 - 1:2000, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_005068.2
Gene Alias E(Sp1) homolog, enhancer of split groucho 1, ESG, ESG1Enhancer of split groucho-like protein 1, GRG1, transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 1, homolog of Drosophila E(sp1), transducin-like enhancer protein 1
Gene Symbols TLE1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 175 and 225 of human Transducin-like Enhancer of Split 1 using the numbering given in entry NP_005068.2 (GeneID 7088).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7088
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.