Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLE1 Antibody (OTI1H2), PerCP, Novus Biologicals™
SDP

Catalog No. NB274530PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274530PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274530PCP Supplier Novus Biologicals Supplier No. NBP274530PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLE1 Monoclonal specifically detects TLE1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen TLE1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1H2
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias E(Sp1) homolog, enhancer of split groucho 1, ESG, ESG1Enhancer of split groucho-like protein 1, GRG1, transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 1, homolog of Drosophila E(sp1), transducin-like enhancer protein 1
Gene Symbols TLE1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 180-460 of human TLE1 (NP_005068) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7088.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.