Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLE1 Rabbit anti-Human, Mouse, Rat, Clone: 6Q5X1, Novus Biologicals™
SDP

Catalog No. NB167508 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167508 20 μg
NB167507 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167508 Supplier Novus Biologicals Supplier No. NBP31620820UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TLE1 Monoclonal antibody specifically detects TLE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen TLE1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6Q5X1
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias E(Sp1) homolog, enhancer of split groucho 1, ESG, ESG1Enhancer of split groucho-like protein 1, GRG1, transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 1, homolog of Drosophila E(sp1), transducin-like enhancer protein 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 150-250 of human TLE1 (Q04724).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7088
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.