Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLE2 Antibody (OTI5A6), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP274531R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274531R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274531R Supplier Novus Biologicals Supplier No. NBP274531R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLE2 Monoclonal specifically detects TLE2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TLE2
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI5A6
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias enhancer of split groucho 2, ESG, ESG2Enhancer of split groucho-like protein 2, FLJ41188, GRG2, transducin-like enhancer of split 2 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 2, homolog of Drosophila E(sp1), transducin-like enhancer protein 2
Gene Symbols TLE2
Host Species Mouse
Immunogen Full length recombinant protein of human TLE2 (NP_003251) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7089.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.