Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR10 Antibody (28M7A3), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP227273C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227273C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227273C Supplier Novus Biologicals Supplier No. NBP227273C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR10 Monoclonal specifically detects TLR10 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TLR10
Applications Flow Cytometry
Classification Monoclonal
Clone 28M7A3
Conjugate DyLight 650
Dilution Flow Cytometry 1ul/1 million cells
Gene Accession No. Q9BXR5
Gene Alias CD290, CD290 antigen, MGC104967, MGC126398, MGC126399, toll-like receptor 10
Gene Symbols TLR10
Host Species Mouse
Immunogen A portion of amino acids 300-350 of human TLR10 was used as immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 81793
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.