Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR10 Antibody (3C10C5), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP227214UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227214UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227214UV Supplier Novus Biologicals Supplier No. NBP227214UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR10 Monoclonal specifically detects TLR10 in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface).

Specifications

Antigen TLR10
Applications Flow Cytometry
Classification Monoclonal
Clone 3C10C5
Conjugate DyLight 350
Dilution Flow Cytometry, Flow (Cell Surface)
Gene Accession No. Q9BXR5
Gene Alias CD290, CD290 antigen, MGC104967, MGC126398, MGC126399, toll-like receptor 10
Gene Symbols TLR10
Host Species Mouse
Immunogen A partial recombinant protein containing amino acids 20-474 (extracellular domain) of human TLR10.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 81793
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.