Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR11 Antibody (786404), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB295710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295710 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295710 Supplier Novus Biologicals Supplier No. FAB7640T

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

TLR11 Monoclonal antibody specifically detects TLR11 in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen TLR11
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 786404
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias hgnc31753, Toll like receptor 11
Host Species Rat
Immunogen E. coli-derived recombinant mouse TLR11, Thr209-Gln325, Accession # Q6R5P0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 442887
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.