Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR12 Antibody (1229C), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB311018 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311018 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311018 Supplier Novus Biologicals Supplier No. FAB8086PECY55
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TLR12 Monoclonal specifically detects TLR12 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen TLR12
Applications Flow Cytometry
Classification Monoclonal
Clone 1229C
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias toll-like receptor 12
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse TLR12, Thr22-Lys709, Accession # Q6QNU9
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131451
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.