Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR12 Antibody (15F1215), Novus Biologicals™
SDP

Catalog No. NBP224870 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224870 0.2 mL
NBP224870SS 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224870 Supplier Novus Biologicals Supplier No. NBP224870
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR12 Monoclonal specifically detects TLR12 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen TLR12
Applications Western Blot
Classification Monoclonal
Clone 15F1215
Conjugate Unconjugated
Dilution Western Blot 1: 1000
Gene Accession No. Q6QNU9
Gene Alias toll-like receptor 12
Gene Symbols TLR12P
Host Species Mouse
Immunogen This antibody was developed against amino acids 791-821 of mouse TLR12; GenBank NP_991392.1
Purification Method Unpurified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131451
Test Specificity This antibody is reactive to Mouse.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.