Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR12 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224833SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224833SS 0.025 mg
NBP224833 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224833SS Supplier Novus Biologicals Supplier No. NBP224833SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

TLR12 Polyclonal specifically detects TLR12 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Flow (Intracellular).

Specifications

Antigen TLR12
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Flow (Intracellular)
Formulation 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide
Gene Alias toll-like receptor 12
Gene Symbols TLR12P
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids of 743-756 (RVWLQGLRGKGDKG) of mouse TLR12.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131451
Target Species Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.