Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR13 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224539 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224539 0.1 mg
NBP224539SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224539 Supplier Novus Biologicals Supplier No. NBP224539
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

TLR13 Polyclonal specifically detects TLR13 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Peptide ELISA, Flow (Intracellular).

Specifications

Antigen TLR13
Applications Western Blot, Flow Cytometry, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Flow Cytometry 1-3 ug/10^6 cells, Peptide ELISA 1:100-1:2000, Flow (Intracellular) reported in scientific literature (PMID 24518205)
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q6R5N8
Gene Alias TLR-13
Gene Symbols TLR13
Host Species Rabbit
Immunogen A portion of amino acid 800 to 850 of mouse TLR13 was used as immunogen for this antibody.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 442889
Test Specificity The amino acid sequence used as immunogen for this p65 NLS antibody is widely conserved in many species and isoforms including human, mouse, rat, rhesus monkey, chimp, bovine and horse.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.