Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR13 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $426.00

Specifications

Antigen TLR13
Concentration 0.5 mg/mL
Dilution Western Blot 1-3 ug/ml, Flow Cytometry 1-3 ug/10^6 cells, Peptide ELISA 1:100-1:2000, Flow (Intracellular) reported in scientific literature (PMID 24518205)
Applications Western Blot, Flow Cytometry, ELISA
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224539SS
SDP
View Documents
Novus Biologicals
NBP224539SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224539
SDP
View Documents
Novus Biologicals
NBP224539
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

TLR13 Polyclonal specifically detects TLR13 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Peptide ELISA, Flow (Intracellular).
Specifications

Specifications

TLR13
Western Blot 1-3 ug/ml, Flow Cytometry 1-3 ug/10^6 cells, Peptide ELISA 1:100-1:2000, Flow (Intracellular) reported in scientific literature (PMID 24518205)
Polyclonal
Rabbit
Mouse, Rat
Q6R5N8
442889
A portion of amino acid 800 to 850 of mouse TLR13 was used as immunogen for this antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Flow Cytometry, ELISA
Unconjugated
RUO
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
TLR-13
TLR13
IgG
Affinity Purified
The amino acid sequence used as immunogen for this p65 NLS antibody is widely conserved in many species and isoforms including human, mouse, rat, rhesus monkey, chimp, bovine and horse.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.