Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR2 Antibody (11G5), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP227166C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227166C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227166C Supplier Novus Biologicals Supplier No. NBP227166C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TLR2 Monoclonal specifically detects TLR2 in Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen TLR2
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 11G5
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry
Gene Alias CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
Gene Symbols TLR2
Host Species Rat
Immunogen A portion of amino acids 400-450 of murine TLR2 was used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 7097
Test Specificity The amino acid sequence used as immunogen is 85% homologous in Vole, 80% in Rat, Chinese Hamster and Rabbit, 70% in Orangutan, Rhesus Monkey and Horse, and 65% homologous in Human.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.