Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR2 Antibody (203325), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB175241 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175241 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175241 Supplier Novus Biologicals Supplier No. FAB1530Y
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

TLR2 Monoclonal antibody specifically detects TLR2 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen TLR2
Applications Flow Cytometry
Classification Monoclonal
Clone 203325
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TLR2, Gln25-Leu590, Accession # Q9QUN7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7097
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.