Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR2 Antibody (67N17F8), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP225244V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225244V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225244V Supplier Novus Biologicals Supplier No. NBP225244V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR2 Monoclonal specifically detects TLR2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TLR2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 67N17F8
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Accession No. NP_003255.2
Gene Alias CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
Gene Symbols TLR2
Host Species Mouse
Immunogen Within the range of amino acids 227-398 of human TLR2 were used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 7097
Test Specificity This product reacts with Human.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.