Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR2 Antibody (67N8F8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP225245SS
Description
TLR2 Monoclonal specifically detects TLR2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface).Specifications
TLR2 | |
Monoclonal | |
0.5 mg/mL | |
Western Blot 1-3 ug/ml, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Cell Surface) | |
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This product reacts with Human. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
67N8F8 | |
Unconjugated | |
NP_003255.2 | |
TLR2 | |
Amino acids 227-371 of human TLR2 were used as the immunogen for the antibody. | |
0.025 mg | |
Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
7097 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction