Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR2 Antibody (67N8F8), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $438.00

Specifications

Antigen TLR2
Clone 67N8F8
Concentration 0.5 mg/mL
Dilution Western Blot 1-3 ug/ml, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Cell Surface)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP225245SS
SDP
View Documents
Novus Biologicals
NBP225245SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP225245
SDP
View Documents
Novus Biologicals
NBP225245
0.1 mg
Each for $438.00
Only null left
Add to Cart
 
Description

Description

TLR2 Monoclonal specifically detects TLR2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface).
Specifications

Specifications

TLR2
0.5 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
NP_003255.2
7097
Amino acids 227-371 of human TLR2 were used as the immunogen for the antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
67N8F8
Western Blot 1-3 ug/ml, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Cell Surface)
Monoclonal
Purified
RUO
Human
CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
TLR2
IgG1 κ
Protein G purified
This product reacts with Human.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.