Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR2 Antibody (TL2.1), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N106722A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106722A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106722A30 Supplier Novus Biologicals Supplier No. NB10056722AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR2 Monoclonal specifically detects TLR2 in Canine, Human, Mouse, Rabbit, Rat samples. It is validated for Block/Neutralize, Dot Blot, ELISA, Flow Cytometry, Flow Cytometry (Cell Surface), Functional Assay, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, In vitro Assay, Western Blot.

Specifications

Antigen TLR2
Applications Blocking Assay, Dot Blot, ELISA, Flow Cytometry, Flow Cytometry (Cell Surface), Functional Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In vitro Assay, Western Blot
Classification Monoclonal
Clone TL2.1
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface)
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2
Gene Symbols TLR2
Host Species Mouse
Immunogen This antibody was raised by immunizing mice with CHO cells transfected with human TLR2 cDNA (Flo et al, 2000). The hybridoma supernatants were selected by flow cytometry.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7097
Target Species Canine, Human, Mouse, Rabbit, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.