Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR3 Antibody (002), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB073745 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB073745 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB073745 Supplier Novus Biologicals Supplier No. NBP290455AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TLR3 Monoclonal specifically detects TLR3 in Mouse samples. It is validated for ELISA.

Specifications

Antigen TLR3
Applications ELISA
Classification Monoclonal
Clone 2
Conjugate Alexa Fluor 700
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD283, CD283 antigen, toll-like receptor 3
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TLR3 (NP_569054.2; Met1-Leu705).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Necroptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7098
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.