Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR3 Antibody (002), PerCP, Novus Biologicals™
SDP

Catalog No. NB333170 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333170 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333170 Supplier Novus Biologicals Supplier No. NBP290455PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TLR3 Monoclonal antibody specifically detects TLR3 in Mouse samples. It is validated for ELISA

Specifications

Antigen TLR3
Applications ELISA
Classification Monoclonal
Clone 002
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias CD283, CD283 antigen, toll-like receptor 3
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse TLR3 (Accession#: NP_569054.2, Met1-Leu705)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Necroptosis, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 7098
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.