Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR3 Antibody (27N3D4), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP227405X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227405X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227405X Supplier Novus Biologicals Supplier No. NBP227405AF488
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TLR3 Monoclonal specifically detects TLR3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen TLR3
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 27N3D4
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry
Gene Alias CD283, CD283 antigen, toll-like receptor 3
Gene Symbols TLR3
Host Species Rat
Immunogen A portion of amino acids 350-375 of mouse TLR3 was used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 7098
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.