Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR3 Antibody (40C1285.6), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP224875W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224875W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224875W Supplier Novus Biologicals Supplier No. NBP224875AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR3 Monoclonal specifically detects TLR3 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen TLR3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 40C1285.6
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular)
Gene Accession No. O15455
Gene Alias CD283, CD283 antigen, toll-like receptor 3
Gene Symbols TLR3
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 55-85 of human TLR3 was used as immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immune System Diseases, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 7098
Test Specificity This antibody is reactive to Dog, Human and Mouse.
Target Species Human, Mouse, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.