Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR4 Antibody (1203B), PerCP, Novus Biologicals™
SDP

Catalog No. NB364329 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364329 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364329 Supplier Novus Biologicals Supplier No. FAB27591C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TLR4 Monoclonal specifically detects TLR4 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen TLR4
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1203B
Conjugate PerCP
Dilution Flow Cytometry, Immunocytochemistry
Formulation PBS
Gene Alias ARMD10, CD_antigen: CD284, CD284, CD284 antigen, EC 3.2.2.6, EC:3.2.2.6, homolog of Drosophila toll, hToll, TLR4, TLR-4, TOLL, toll like receptor 4 protein, toll-like receptor 4
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TLR4, Asn26-Lys629, Accession # Q9QUK6
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Autophagy, Cytokine Research, Immunology, Inflammation, Innate Immunity, Neurodegeneration, Signal Transduction, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 7099
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.