Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR4 Antibody (76B357.1), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP227149X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227149X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227149X Supplier Novus Biologicals Supplier No. NBP227149AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR4 Monoclonal specifically detects TLR4 in Human, Mouse, Rat, Porcine, Bovine, Mammal samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface), Knockout Validated, Knockdown Validated.

Specifications

Antigen TLR4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown
Classification Monoclonal
Clone 76B357.1
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface), Knockout Validated, KnockDown Validated
Gene Alias ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4
Gene Symbols TLR4
Host Species Mouse
Immunogen This antibody was developed against a portion of amino acids 100-200 of human TLR4 (NP_612564).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 7099
Test Specificity The amino acid sequence used has 100% identity in human, chimp, baboon, Chinese hamster, and rat; 93% identity in bovine and pig; 85% identity in sheep and 78% identity in cat, horse and mouse.
Target Species Human, Mouse, Rat, Pig, Bovine, Mammalia
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.