Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR4 Antibody (76B357.1), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N105656APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N105656APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N105656APC7 Supplier Novus Biologicals Supplier No. NB10056566APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR4 Monoclonal specifically detects TLR4 in Bovine, Human, Mammalia, Mouse, Pig, Rat samples. It is validated for Block/Neutralize, ChIP Assay, CyTOF, Dot Blot, Dual RNAScope ISH-IHC, ELISA, ELISA Capture (Matched Antibody Pair), Flow Cytometry, Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In vitro Assay, KnockDown, SDS-Page, Western Blot.

Specifications

Antigen TLR4
Applications Blocking Assay, ChIP Assay, CyTOF, Dot Blot, Dual RNAScope ISH-IHC, ELISA, ELISA Capture (Matched Antibody Pair), Flow Cytometry, Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In vitro Assay, KnockDown, SDS-Page, Western Blot
Classification Monoclonal
Clone 76B357.1
Conjugate APC-Cyanine7
Dilution Flow Cytometry, Knockout Validated, KnockDown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4
Gene Symbols TLR4
Host Species Mouse
Immunogen This antibody was developed against a portion of amino acids 100-200 of human TLR4 (NP_612564).
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 7099.0
Target Species Bovine, Human, Mammalia, Mouse, Pig, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.