Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR4 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 12 publications

$192.47 - $529.95

Specifications

Antigen TLR4
Concentration 1.12 mg/mL
Dilution Western Blot 1 - 3 ug/mL, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:25 - 1:75, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen reported in scientific literature (PMID 24128422), Immunoblotting reported in scientific literature (PMID 23744481), Proximity Ligation Assay reported in scientific literature (PMID 30579042)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056581T
SDP
View Documents
Novus Biologicals
NB10056581SS
0.025 mL
Each for $192.47
Only null left
Add to Cart
 
NB10056581
SDP
View Documents
Novus Biologicals
NB10056581
0.1 mL
Each for $529.95
Only null left
Add to Cart
 
Description

Description

TLR4 Polyclonal specifically detects TLR4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Proximity Ligation Assay.
Specifications

Specifications

TLR4
Western Blot 1 - 3 ug/mL, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:25 - 1:75, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen reported in scientific literature (PMID 24128422), Immunoblotting reported in scientific literature (PMID 23744481), Proximity Ligation Assay reported in scientific literature (PMID 30579042)
Polyclonal
Rabbit
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
PBS and 30% Glycerol with 0.05% Sodium Azide
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4
TLR4
IgG
Affinity Purified
75 kDa
1.12 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat
Q9QUK6
7099
This TLR4 antibody was developed against a synthetic peptide made to an internal portion of the mouse TLR4 protein (between residues 400-450) [Q9QUK6].
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.