Learn More
Description
Specifications
Specifications
| Antigen | TLR4 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.12 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 - 3 ug/mL, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:25 - 1:75, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen reported in scientific literature (PMID 24128422), Immunoblotting reported in scientific literature (PMID 23744481), Proximity Ligation Assay reported in scientific literature (PMID 30579042) |
| Formulation | PBS and 30% Glycerol with 0.05% Sodium Azide |
| Gene Accession No. | Q9QUK6 |
| Gene Alias | ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 |
| Gene Symbols | TLR4 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
