Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR4 Antibody (HTA125) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB156727SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB156727SS 0.025 mg
NB10056727 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB156727SS Supplier Novus Biologicals Supplier No. NB10056727SS

Mouse Monoclonal Antibody has been used in 26 publications

TLR4 Monoclonal specifically detects TLR4 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunoblotting, In vitro assay, Flow (Cell Surface), Flow (Intracellular), CyTOF-ready, Block/Neutralize.

Specifications

Antigen TLR4
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoblot, In vitro Assay
Classification Monoclonal
Clone HTA125
Conjugate Unconjugated
Dilution Western Blot reported in scientific literature (PMID 28546218), Flow Cytometry 1ul/1 million cells, Immunocytochemistry/Immunofluorescence 1:10 - 1:2000. Use reported in scientific literature (Schneeman et al (2005)), Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (Shimazu et al (1999)), Immunoblotting reported in scientific literature (PMID 19195606), In vitro assay reported in scientific literature (PMID 24512642), Flow (Cell Surface) 2-5 ug/1 million cells, Flow (Intracellular), CyTOF-ready, Block/Neutralize reported in scientific literature (PMID 24512642)
Formulation Filter (0.2 micron) sterilized PBS with No Preservative
Gene Accession No. O00206
Gene Alias ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4
Gene Symbols TLR4
Host Species Mouse
Immunogen This antibody was developed by immunizing mice with Ba/F3 cell line expressing human TLR4 cell surface antigen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7099
Test Specificity The has been shown to block the activation of monocytes with LPS (Paik, et al, 2003).
Target Species Human, Mouse, Canine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.