Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR4 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB337408 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337408 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337408 Supplier Novus Biologicals Supplier No. NBP224538MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TLR4 Polyclonal antibody specifically detects TLR4 in Human, Mouse, Bovine, Equine, Ferret, Primate, Macaque samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen TLR4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ARMD10, CD_antigen: CD284, CD284, CD284 antigen, EC 3.2.2.6, EC:3.2.2.6, homolog of Drosophila toll, hToll, TLR4, TLR-4, TOLL, toll like receptor 4 protein, toll-like receptor 4
Host Species Rabbit
Immunogen This TLR4 antibody was developed against a portion of amino acids 650-700 of human TLR4.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Autophagy, Cytokine Research, Immunology, Inflammation, Innate Immunity, Neurodegeneration, Signal Transduction, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 7099
Target Species Human, Mouse, Bovine, Equine, Ferret
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.