Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR4 Antibody (MTS510), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP224865FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224865FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224865FR Supplier Novus Biologicals Supplier No. NBP224865FR
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TLR4 Monoclonal specifically detects TLR4 in Mouse samples. It is validated for Flow Cytometry, Flow (Cell Surface).

Specifications

Antigen TLR4
Applications Flow Cytometry
Classification Monoclonal
Clone MTS510
Conjugate DyLight 680
Dilution Flow Cytometry, Flow (Cell Surface)
Gene Accession No. O00206
Gene Alias ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4
Gene Symbols TLR4
Host Species Rat
Immunogen This antibody was developed by immunizing rats with the Ba/F3 cell line expressing mouse TLR4 and MD-2 (Akashi et al, 2000).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 7099
Test Specificity This antibody preferentially recognizes TLR4-MD-2 complex than of TLR4 alone.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.