Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR4 Antibody (TLR4/3895R), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB391093 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391093 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391093 Supplier Novus Biologicals Supplier No. NBP308732AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TLR4 Monoclonal antibody specifically detects TLR4 in Human, Rat, Porcine, Canine, Guinea Pig, Monkey samples. It is validated for ELISA

Specifications

Antigen TLR4
Applications ELISA
Classification Monoclonal
Clone TLR4/3895R
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias ARMD10, CD_antigen: CD284, CD284, CD284 antigen, EC 3.2.2.6, EC:3.2.2.6, homolog of Drosophila toll, hToll, TLR4, TLR-4, TOLL, toll like receptor 4 protein, toll-like receptor 4
Host Species Rabbit
Immunogen Recombinant full-length human TLR4 protein. (Uniprot: O00206)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Autophagy, Cytokine Research, Immunology, Inflammation, Innate Immunity, Neurodegeneration, Signal Transduction, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 7099
Target Species Human, Rat, Pig, Canine, Guinea Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.