Learn More
TLR4/MD-2 Complex Monoclonal Antibody (MTS510), Super Bright™ 436, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
$476.00
Specifications
| Antigen | TLR4/MD-2 Complex |
|---|---|
| Clone | MTS510 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
62-992-482
![]()
|
Invitrogen™
62992482 |
100 μg |
Each of 1 for $476.00
|
|
|||||
Description
The MTS510 monoclonal antibody reacts with the mouse Toll-like receptor 4 (TLR4)/MD-2 complex. At least ten members of the Toll family have been identified. This family of type I transmembrane proteins is characterized by an extracellular domain with leuc
MD-2 is a member of the Toll/interleukin-1 receptor (TIR) family, a group of proteins that include the Toll-like receptors (TLRs). TLRs are signaling molecules that recognize different pathogen-associated molecular patterns (PAMPs) and serve as an important link between the innate and adaptive immune responses. TLR4, the major signaling receptor for lipopolysaccharide (LPS), requires the binding of MD-2 to its extracellular region for maximal response to LPS. The specificity of this response is determined by the species of MD-2; e.g., human MD-2 transfected into mouse cells can cause mouse TLR4 to react to LPS analogs that are normally antagonistic to human but not mouse TLR4.Specifications
| TLR4/MD-2 Complex | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Q9JHF9, Q9QUK6 | |
| Ly96, Tlr4 | |
| Primary | |
| Antibody |
| MTS510 | |
| Flow Cytometry | |
| Super Bright 436 | |
| Rat | |
| Mouse | |
| 17087, 21898 | |
| IgG2a κ | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Ly96 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.