Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR5 Antibody (624915), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB311545 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311545 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311545 Supplier Novus Biologicals Supplier No. FAB6704PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR5 Monoclonal antibody specifically detects TLR5 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen TLR5
Applications Flow Cytometry
Classification Monoclonal
Clone 624915
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5
Host Species Mouse
Immunogen E. coli-derived recombinant human TLR5, Ile21-Phe115, Accession # O60602
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7100
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.