Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR5 Antibody (85B152.5), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP197728UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197728UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197728UV Supplier Novus Biologicals Supplier No. NBP197728UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR5 Monoclonal specifically detects TLR5 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular).

Specifications

Antigen TLR5
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 85B152.5
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular)
Gene Accession No. O60602
Gene Alias MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5
Gene Symbols TLR5
Host Species Mouse
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5. It will cross-react with mouse TLR5 (NP_003259).
Molecular Weight of Antigen 100 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 7100
Target Species Human, Mouse, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.