Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224827SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224827SS 0.025 mg
NBP224827 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224827SS Supplier Novus Biologicals Supplier No. NBP224827SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 8 publications

TLR5 Polyclonal specifically detects TLR5 in Human, Mouse, Pig, Rat samples. It is validated for Flow Cytometry, Flow Cytometry (Cell Surface), IF/IHC, Immunohistochemistry, Immunohistochemistry (Paraffin), In vitro Assay, Western Blot.

Specifications

Antigen TLR5
Applications Flow Cytometry, Flow Cytometry (Cell Surface), Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), In vitro Assay, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/mL, Flow Cytometry 1 - 2 ug/10^6 cells, Immunohistochemistry reported in scientific literature (PMID 30716659), Immunohistochemistry-Paraffin, In vitro assay reported in scientific literature (PMID 26084721), Flow (Cell Surface) reported in scientific literature (PMID 24532644)
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. O60602
Gene Alias MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5
Gene Symbols TLR5
Host Species Rabbit
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5 found between amino acids 300-350.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 7100
Test Specificity This antibody is reactive to Human, Mouse and Rat. Porcine reactivity reported in scientific literature (PMID: 24097563).
Target Species Human, Mouse, Pig, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.