Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR5 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 8 publications

$206.50 - $501.50

Specifications

Antigen TLR5
Concentration 1.0 mg/mL
Dilution Western Blot 1 - 3 ug/mL, Flow Cytometry 1 - 2 ug/10^6 cells, Immunohistochemistry reported in scientific literature (PMID 30716659), Immunohistochemistry-Paraffin, In vitro assay reported in scientific literature (PMID 26084721), Flow (Cell Surface) reported in scientific literature (PMID 24532644)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), In vitro Assay
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224827SS
SDP
View Documents
Novus Biologicals
NBP224827SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224827
SDP
View Documents
Novus Biologicals
NBP224827
0.1 mg
Each for $501.50
Only null left
Add to Cart
 
Description

Description

TLR5 Polyclonal specifically detects TLR5 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, In vitro assay, Flow (Cell Surface).
Specifications

Specifications

TLR5
Western Blot 1 - 3 ug/mL, Flow Cytometry 1 - 2 ug/10^6 cells, Immunohistochemistry reported in scientific literature (PMID 30716659), Immunohistochemistry-Paraffin, In vitro assay reported in scientific literature (PMID 26084721), Flow (Cell Surface) reported in scientific literature (PMID 24532644)
Polyclonal
Purified
RUO
Human, Mouse, Rat, Pig
O60602
7100
This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TLR5 found between amino acids 300-350.
Primary
Store at -20C. Avoid freeze-thaw cycles.
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), In vitro Assay
Unconjugated
Rabbit
Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors
PBS with 0.05% Sodium Azide
MGC126430, MGC126431, SLEB1FLJ10052, TIL3Toll/interleukin-1 receptor-like protein 3, toll-like receptor 5
TLR5
IgG
Protein G purified
This antibody is reactive to Human, Mouse and Rat. Porcine reactivity reported in scientific literature (PMID: 24097563).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.