Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR6 Antibody (418601), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB349852 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349852 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349852 Supplier Novus Biologicals Supplier No. FAB1533MFV500
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TLR6 Monoclonal specifically detects TLR6 in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen TLR6
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 418601
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD286, CD286 antigen, toll-like receptor 6
Host Species Rat
Immunogen HEK293 human embryonic kidney cell line transfected with mouse TLR6, Phe39-Thr806, Accession # BAA78632
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10333
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.