Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR6 Antibody (86B1153.2), Biotin, Novus Biologicals™
SDP

Catalog No. NBP224791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224791 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224791 Supplier Novus Biologicals Supplier No. NBP224791
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 9 publications

TLR6 Monoclonal specifically detects TLR6 in Human samples. It is validated for Flow Cytometry, ELISA, Flow (Cell Surface).

Specifications

Antigen TLR6
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 86B1153.2
Concentration 0.5mg/mL
Conjugate Biotin
Dilution Flow Cytometry, ELISA 1:100-1:2000, Flow (Cell Surface)
Gene Accession No. Q9Y2C9
Gene Alias CD286, CD286 antigen, toll-like receptor 6
Gene Symbols TLR6
Host Species Mouse
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 408-424 of human TRL6.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 10333
Test Specificity TLR6 (86B1153.2)
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.