Learn More
Description
Specifications
Specifications
| Antigen | TLR6 |
| Applications | Blocking Assay, Flow Cytometry, Flow Cytometry (Cell Surface), Flow Cytometry (Intracellular Staining), Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 86B1153.2 |
| Concentration | 1.0 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot reported in scientific literature (PMID 17332440), Flow Cytometry 3 ug/10-6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Cell Surface) reported in scientific literature (PMID 24986635), Flow (Intracellular) |
| Gene Accession No. | Q9Y2C9 |
| Gene Alias | CD286, CD286 antigen, toll-like receptor 6 |
| Gene Symbols | TLR6 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
