Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR6 Antibody (86B1153.2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 17 publications
$206.50 - $499.50
Specifications
Antigen | TLR6 |
---|---|
Clone | 86B1153.2 |
Concentration | 1.0 mg/ml |
Dilution | Western Blot reported in scientific literature (PMID 17332440), Flow Cytometry 3 ug/10-6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Cell Surface) reported in scientific literature (PMID 24986635), Flow (Intracellular) |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056536T
![]() |
Novus Biologicals
NB10056536SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056536
![]() |
Novus Biologicals
NB10056536 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
TLR6 Monoclonal specifically detects TLR6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface), Flow (Intracellular).Specifications
TLR6 | |
1.0 mg/ml | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Immunology, Innate Immunity, Prostate Cancer, Toll Like Receptors | |
Q9Y2C9 | |
10333 | |
This antibody was developed against a synthetic peptide corresponding to amino acids 408-424 of human TRL6. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
86B1153.2 | |
Western Blot reported in scientific literature (PMID 17332440), Flow Cytometry 3 ug/10-6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Cell Surface) reported in scientific literature (PMID 24986635), Flow (Intracellular) | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD286, CD286 antigen, toll-like receptor 6 | |
TLR6 | |
IgG1 | |
Protein G purified | |
TLR6 (86B1153.2) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title