Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR6 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB198076 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB198076 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB198076 Supplier Novus Biologicals Supplier No. NBP176664AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TLR6 Polyclonal antibody specifically detects TLR6 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen TLR6
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias CD286, CD286 antigen, toll-like receptor 6
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 13 amino acids near the center of human TLR6. The immunogen is located within amino acids 390 - 440 of TLR6. Amino Acid Squence: SWNSLESGRHKEN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10333
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.