Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR7 Antibody (4F4), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP104349A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP104349A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP104349A30 Supplier Novus Biologicals Supplier No. NBP104349AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR7 Monoclonal specifically detects TLR7 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TLR7
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 4F4
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias toll-like receptor 7
Gene Symbols TLR7
Host Species Mouse
Immunogen Recombinant human TLR7 (451-500aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 51284.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.