Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLR7 Antibody (4F4), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N104349PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N104349PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N104349PEC7 Supplier Novus Biologicals Supplier No. NBP104349PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TLR7 Monoclonal specifically detects TLR7 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TLR7
Applications Flow Cytometry
Classification Monoclonal
Clone 4F4
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias toll-like receptor 7
Gene Symbols TLR7
Host Species Mouse
Immunogen Recombinant human TLR7 (451-500aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 51284.0
Test Specificity Anti-human TLR7 mAb, clone 4F4, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human TLR7 protein.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.